Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA3 Monoclonal Antibody (8A3C10D11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530852
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SerpinA3/SERPINA3.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
Specifications
SERPINA3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01011 | |
SERPINA3 | |
Recombinant Human SerpinA3 protein (Met1-Ala423). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
8A3C10D11 | |
Unconjugated | |
SERPINA3 | |
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
Mouse | |
Protein A | |
RUO | |
12 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction