Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587710 Supplier Invitrogen™ Supplier No. PA587710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPNSPLDEEN LTQENQDRGT HVDLGLASAN VDFAFSLYKQ LVLKAPDKNV IFSPLSISTA LAFLSLGAHN TTLTEILKGL KFNLTETSEA EIHQSFQHLL RTLNQSSDEL QLSMGNAMFV KEQLSLLDRF TEDAKRLYGS EAFATDFQDS AAAKKLINDY VKNGTRGKIT DLIKDLDSQT MMVLVNYIFF KAKWEMPFDP QDTHQSRFYL SKKKWVMVPM MSLHHLTIPY FRDEELSCTV VELKYTGNAS ALFILPDQDK MEEVEAMLLP ETLKRWRDSL EFREIGELYL PKFSISRDYN LNDILLQLGI EEAFTSKADL SGITGARNLA VSQVVHKAVL DVFEEGTEAS AATAVKITLL SALVETRTIV RFNRPFLMII VPTDTQNIFF MSKVTNPKQA; Positive Samples: Mouse testis, Mouse liver, Rat liver.

SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SERPINA3
Gene Accession No. P01011
Gene Alias AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3
Gene Symbols SERPINA3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.