Learn More
Invitrogen™ SERPINA3 Polyclonal Antibody

Description
PA1-4150 detects the ACT protein in human samples. PA1-4150 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~33 kDa protein representing the ACT protein in testis extract. The PA1- 4150 immunogen is the N-terminus of the human ACT protein.
Specifications
Specifications
| Antigen | SERPINA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SERPINA3 |
| Gene Accession No. | P01011 |
| Gene Alias | AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 |
| Gene Symbols | SERPINA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.