Learn More
Invitrogen™ SERPINA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120834
Description
Positive test controls include: HepG2, Human plasma, Mouse uterus, Rat uterus. The target is usually found in the following locations: Secreted. Immunogen sequence: HPNSPLDEEN LTQENQDRGT HVDLGLASAN VDFAFSLYKQ LVLKAPDKNV IFSPLSISTA LAFLSLGAHN TTLTEILKGL KFNLTETSEA EIHQSFQHLL RTLNQSSDEL QLSMGNAMFV KEQLSLLDRF TEDAKRLYGS EAFATDFQDS AAAKKLINDY VKNGTRGKIT DLIKDLDSQT MMVLVNYIFF KAKWEMPFDP QDTHQSRFYL SKKKWVMVPM MSLHHLTIPY FRDEELSCTV VELKYTGNAS ALFILPDQDK MEEVEAMLLP ETLKRWRDSL EFREIGELYL PKFSISRDYN LNDILLQLGI EEAFTSKADL SGITGARNLA VSQVVHKAVL DVFEEGTEAS AATAVKITLL SALVETRTIV RFNRPFLMII VPTDTQNIFF MSKVTNPKQA.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
Specifications
SERPINA3 | |
Polyclonal | |
Unconjugated | |
SERPINA3 | |
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P01011 | |
SERPINA3 | |
A synthetic peptide corresponding to a sequence within amino acids 24-423 of human Alpha-1 Antichymotrypsin (SERPINA3) (NP_0010762). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.