Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547180 Supplier Thermo Scientific Supplier No. PA547180
Add to Cart
Add to Cart

SERPINA4 Polyclonal Antibody

SERPINA4 Polyclonal Antibody for Western Blot, IHC, IP, Neu

TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA4
Applications Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SERPINA4
Gene Accession No. P29622
Gene Alias KAL; Kallikrein inhibitor; kallistatin; KLST; KST; Peptidase inhibitor 4; PI4; PI-4; protease inhibitor 4 (kallistatin); serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; serpin A4; serpin family A member 4; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; SERPINA4; sserpin family A member 4; testicular secretory protein Li 22
Gene Symbols SERPINA4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Serpin A4/Kallistatin Gly20-Pro427 (predicted).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5267
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.