Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582109 Supplier Invitrogen™ Supplier No. PA582109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLTELSESDV HRGFQHLLHT LNLPGHGLET RVGSALFLSH NLKFLAKFLN DTMAVYEAKL FHTNFYDTVG TIQLINDHVK KETRGKIVDL VSELKKDVLM VLVNYIYFKA LWEKPFISSR TTPKDFYVDE NTTVRVPMML QDQE Highest antigen sequence indentity to the following orthologs: Mouse - 47%, Rat - 53%.

SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SERPINA4
Gene Accession No. P29622
Gene Alias KAL; Kallikrein inhibitor; kallistatin; KLST; KST; Peptidase inhibitor 4; PI4; PI-4; protease inhibitor 4 (kallistatin); serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; serpin A4; serpin family A member 4; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; SERPINA4; sserpin family A member 4; testicular secretory protein Li 22
Gene Symbols SERPINA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERPINA4. Recombinant protein control fragment (Product #RP-89034).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.