Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA4 Recombinant Rabbit Monoclonal Antibody (6D0O7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551289
Description
Immunogen sequence: MHLIDYLLLL LVGLLALSHG QLHVEHDGES CSNSSHQQIL ETGEGSPSLK IAPANADFAF RFYYLIASET PGKNIFFSPL SISAAYAMLS LGACSHSRSQ.
SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.
Specifications
SERPINA4 | |
Recombinant Monoclonal | |
0.09 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
KAL; Kallikrein inhibitor; kallistatin; KLST; KST; Peptidase inhibitor 4; PI4; PI-4; protease inhibitor 4 (kallistatin); serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; serpin A4; serpin family A member 4; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; SERPINA4; sserpin family A member 4; testicular secretory protein Li 22 | |
Rabbit | |
Affinity chromatography | |
RUO | |
246328 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
ELISA, Western Blot | |
6D0O7 | |
Unconjugated | |
SERPINA4 | |
SERPINA4 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Kallistatin (SERPINA4) (NP_006206.2). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction