Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA5 Monoclonal Antibody (OTI1A12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525890
Description
SERPINA5 Monoclonal Antibody for Western Blot
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
SERPINA5 | |
Monoclonal | |
0.54 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05154, P70458 | |
SERPINA5 | |
Full length human recombinant protein of SERPINA5 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Canine | |
Antibody | |
IgG1 |
Western Blot | |
OTI1A12 | |
Unconjugated | |
SERPINA5 | |
4933415L04; acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; LOC480424; LOC483954; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin B3; serpin family A member 5; serpin family B member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINA5; SERPINB3; SERPINB4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
268591, 5104 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction