Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA5 Monoclonal Antibody (OTI1F8), TrueMAB™, OriGene
SDP

Catalog No. 502540359
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0359 30 μL
50-167-5085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0359 Supplier OriGene Supplier No. TA504371S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINA5 Monoclonal antibody specifically detects SERPINA5 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen SERPINA5
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINA5
Gene Accession No. P05154
Gene Alias 4933415L04; acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; LOC480424; LOC483954; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin B3; serpin family A member 5; serpin family B member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINA5; SERPINB3; SERPINB4
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINA5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5104
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.