Learn More
SERPINA5 Mouse anti-Human, Clone: OTI1E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504480

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
SERPINA5 | |
Monoclonal | |
Unconjugated | |
SERPINA5 | |
PAI-3, PAI3, PCI, PCI-B, PLANH3, PROCI | |
Mouse | |
Affinity Chromatography | |
RUO | |
5104 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05154 | |
SERPINA5 | |
Full length human recombit protein of human SERPINA5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.