Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA6 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142692
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Specifications
SERPINA6 | |
Polyclonal | |
Unconjugated | |
SERPINA6 | |
AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
299270 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P31211 | |
SERPINA6 | |
Peptide with sequence C-KASQQINQHVKDKT. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction