Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA6 Polyclonal Antibody

Catalog No. PIPA5121620
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121620 20 μL
PIPA5119053 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121620 Supplier Invitrogen™ Supplier No. PA5121620
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SERPINA6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SERPINA6
Gene Accession No. P08185
Gene Alias AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin
Gene Symbols SERPINA6
Host Species Rabbit
Immunogen Recombinant protein Corticosteroid Binding Globulin. The antigen corresponds to amino acid range 280-405 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 866
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.