Learn More
Invitrogen™ SERPINA6 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547568
Description
In direct ELISAs, approximately 25% cross-reactivity with recombinant rat Serpin A6 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin A1 and rhSerpin A5 is observed. In Western blots, less than 5% cross-reactivity with recombinant human Serpin A4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Specifications
SERPINA6 | |
Polyclonal | |
Unconjugated | |
SERPINA6 | |
AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12401 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q06770 | |
SERPINA6 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin A6 Val23-Ala397. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.