Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA6 Recombinant Rabbit Monoclonal Antibody (SAIC-42B-10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547922
Description
Specificity: Recognises human SERPINA6.
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Specifications
SERPINA6 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P08185 | |
SERPINA6 | |
Peptide AQLLQGLGFNLTER derived from the Corticosteroid-binding globulin SERPINA6, conjugated to KLH. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
SAIC-42B-10 | |
Unconjugated | |
SERPINA6 | |
AI265318; AV104445; Cbg; corticosteroid binding globulin; corticosteroid-binding globulin; serine (or cysteine) peptidase inhibitor, clade A, member 6; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serine (or cysteine) proteinase inhibitor, clade A, member 6; serpin A6; serpin family A member 6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; serpin peptidase inhibitor, clade A, member 6; Serpina6; Transcortin | |
Rabbit | |
Protein A | |
RUO | |
866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction