Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA7 Monoclonal Antibody (1C3H11)
GREENER_CHOICE

Catalog No. FERMA548603
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548603 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548603 Supplier Invitrogen™ Supplier No. MA548603
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINA7 Monoclonal Antibody for Western Blot, ELISA

There are three proteins including thyroxine-binding globulin (TBG), transthyretin and albumin responsible for carrying the thyroid hormones thyroxine (T4) and 3,5,3'-triiodothyronine (T3) in the bloodstream. This gene encodes the major thyroid hormone transport protein, TBG, in serum. It belongs to the serpin family in genomics, but the protein has no inhibitory function like many other members of the serpin family. Mutations in this gene result in TGB deficiency, which has been classified as partial deficiency, complete deficiency, and excess, based on the level of serum TBG. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C3H11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Serpina7
Gene Accession No. P05543
Gene Alias C730040N12Rik; RP1-82J11.2; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antipeptidase, antitrypsin), member 7; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; serine (or cysteine) proteinase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 7; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; serpin A7; serpin family A member 7; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; Serpina7; T4-binding globulin; TBG; thyroxin-binding globulin; thyroxine binding globulin; thyroxine-binding globulin
Gene Symbols Serpina7
Host Species Mouse
Immunogen Purified recombinant fragment of human SERPINA7 (AA: 168-302) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.