Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                SERPINA7 Mouse anti-Human, Mouse, Clone: 1B11B10, Proteintech
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | SERPINA7 | 
|---|---|
| Clone | 1B11B10 | 
| Concentration | 1 mg/mL | 
| Applications | Immunohistochemistry (Paraffin), Western Blot | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        50-173-6880
                    
                    
                        
                    
                        
                    
                    
                        ![]()  
             | 
            
                
                    
                        Proteintech Group Inc
                         664541IG150UL  | 
                
            
            
            
            
            
                
                    150 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $518.60
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
There are three proteins including thyroxine-binding globulin (TBG), transthyretin and albumin responsible for carrying the thyroid hormones thyroxine (T4) and 3,5,3′-triiodothyronine (T3) in the bloodstream. This gene encodes the major thyroid hormone transport protein, TBG, in serum. It belongs to the serpin family in genomics, but the protein has no inhibitory function like many other members of the serpin family. Mutations in this gene result in TGB deficiency, which has been classified as partial deficiency, complete deficiency, and excess, based on the level of serum TBG. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.Specifications
| SERPINA7 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Serpin A7, SERPINA7, T4 binding globulin, TBG, Thyroxine binding globulin | |
| Serpina7 | |
| IgG2a | |
| Protein A | |
| Antibody | 
| 1B11B10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P05543, P61939 | |
| 331535, 6906 | |
| SERPINA7 Fusion Protein ag9823 | |
| Primary | |
| -20°C | |
| SERPINA7 | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title