Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA9 Monoclonal Antibody (136CT29.7.4)
GREENER_CHOICE

Catalog No. PIMA537472
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537472 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537472 Supplier Invitrogen™ Supplier No. MA537472
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINA9 Monoclonal Antibody for Western Blot

SERPINA9 is a protease inhibitor that inhibits trypsin and trypsin-like serine proteases (in vitro). This protein inhibits plasmin and thrombin with lower efficiency (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA9
Applications Western Blot
Classification Monoclonal
Clone 136CT29.7.4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SERPINA9
Gene Accession No. Q86WD7
Gene Alias Centerin; GCET1; Germinal center B-cell-expressed transcript 1 protein; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; serine proteinase inhibitor A11; serpin A9; serpin family A member 9; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; SERPINA11; SERPINA11b; SERPINA9; UNQ692/PRO1337
Gene Symbols SERPINA9
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 158-186 amino acids from the N-terminal region of human SPA9.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.