Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA9 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550618

Catalog No. PA550618


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total Serpin A9 / Centerin protein.

SERPINA9 is a protease inhibitor that inhibits trypsin and trypsin-like serine proteases (in vitro). This protein inhibits plasmin and thrombin with lower efficiency (in vitro).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SERPINA9
Polyclonal
Unconjugated
SERPINA9
Centerin; GCET1; Germinal center B-cell-expressed transcript 1 protein; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; serine proteinase inhibitor A11; serpin A9; serpin family A member 9; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; SERPINA11; SERPINA11b; SERPINA9; UNQ692/PRO1337
Rabbit
Antigen affinity chromatography
RUO
327657
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
2.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86WD7
SERPINA9
Synthetic peptide corresponding to a region derived from internal residues of human serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.