Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB1 Antibody (04), Alexa Fluor™ 350 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305846AF350
Description
SERPINB1 Monoclonal antibody specifically detects SERPINB1 in Human samples. It is validated for ELISASpecifications
SERPINB1 | |
Monoclonal | |
Alexa Fluor 350 | |
EILEI, ELANH2MNEI, elastase, leukocyte elastase inhibitor, Monocyte/neutrophil elastase inhibitor, Peptidase inhibitor 2, PI-2, PI2M/NEI, protease inhibitor 2 (elastase), monocyte/neutrophil derived, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1, Serpin B1, serpin peptidase inhibitor, clade B (ovalbumin), member 1 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SERPINB1 (P30740; Met 1-Pro 379). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
04 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
1992 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction