Learn More
Invitrogen™ SERPINB1 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SERPINB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SERPINB1 |
| Gene Accession No. | P30740 |
| Gene Alias | 1190005M04Rik; AI325983; Anti elastase; anti-elastase; EGK_14416; EI; EIA; ELANH2; epididymis luminal protein 57; epididymis secretory protein Li 27; HEL57; HEL-S-27; L M/N MN PI2; LEI; leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; M/NEI; MNEI; Monocyte/neutrophil elastase inhibitor; ovalbumin; peptidase inhibitor 2; PI2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) peptidase inhibitor, clade B, member 1a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1a; serine (or cysteine) proteinase inhibitor, clade B, member 1a; serine protease inhibitor EIA; serpin; Serpin B1; Serpin B1a; serpin family B member 1; serpin peptidase inhibitor, clade B (ovalbumin), member 1; Serpinb1; Serpinb1a |
| Gene Symbols | SERPINB1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.