Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB10 Monoclonal Antibody (2B8)
GREENER_CHOICE

Catalog No. PIMA524419
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524419 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524419 Supplier Invitrogen™ Supplier No. MA524419
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: AKGTTAAQMA QVLQFNRDQG VKCDPESEKK RKMEFNLSNS EEIHSDFQTL ISEILKPNDD YLLKTANAIY GEKTYAFHNK YLEDMKTYFG AEPQPVNFVE.

Protease inhibitor that may play a role in the regulation of protease activities during hematopoiesis and apoptosis induced by TNF. May regulate protease activities in the cytoplasm and in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB10
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SERPINB10
Gene Accession No. P48595
Gene Alias 9830131G07; BB233602; Bomapin; peptidase inhibitor 10; PI10; PI-10; protease inhibitor 10 (ovalbumin type, bomapin); serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10, pseudogene; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10; serpin B10; serpin family B member 10; serpin peptidase inhibitor, clade B (ovalbumin), member 10; SERPINB10; Serpinb10-ps; TGF-beta-repressible serine proteinase inhibitor; transforming growth factor beta repressible serpin; Trespin
Gene Symbols SERPINB10
Host Species Mouse
Immunogen SERPINB10 (NP_005015.1, 46 a.a. ~ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5273
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.