Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104132 Supplier Invitrogen™ Supplier No. PA5104132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SERPINB10.

Protease inhibitor that may play a role in the regulation of protease activities during hematopoiesis and apoptosis induced by TNF. May regulate protease activities in the cytoplasm and in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERPINB10
Gene Accession No. P48595, Q8K1K6, Q8K3K4
Gene Alias 9830131G07; BB233602; Bomapin; peptidase inhibitor 10; PI10; PI-10; protease inhibitor 10 (ovalbumin type, bomapin); serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10, pseudogene; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10; serpin B10; serpin family B member 10; serpin peptidase inhibitor, clade B (ovalbumin), member 10; SERPINB10; Serpinb10-ps; TGF-beta-repressible serine proteinase inhibitor; transforming growth factor beta repressible serpin; Trespin
Gene Symbols SERPINB10
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERPINB10(Accession P48595), corresponding to amino acid residues I182-E232.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241197, 266775, 5273
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.