Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB10 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278177
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8177 Supplier Leinco Technologies Inc Supplier No. 56312100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SERPINB10 Polyclonal antibody specifically detects SERPINB10 in Human samples. It is validated for Western Blot

Specifications

Antigen SERPINB10
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SERPINB10
Gene Accession No. P48595
Gene Alias 9830131G07; BB233602; Bomapin; peptidase inhibitor 10; PI10; PI-10; protease inhibitor 10 (ovalbumin type, bomapin); serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10, pseudogene; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10; serpin B10; serpin family B member 10; serpin peptidase inhibitor, clade B (ovalbumin), member 10; SERPINB10; Serpinb10-ps; TGF-beta-repressible serine proteinase inhibitor; transforming growth factor beta repressible serpin; Trespin
Host Species Rabbit
Immunogen Peptide corresponding to aa 316-330 of human SERPINB10 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5273
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.