Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522813
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522813 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522813 Supplier Invitrogen™ Supplier No. PA522813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERPINB12 Polyclonal Antibody for Western Blot

Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. A novel member of the human ov-serpin gene family, SERPINB12 is expressed in many tissues, including brain, bone marrow, lymph node, heart, lung, liver, pancreas, testis, ovary, and intestines. SERPINB12 is an inhibitor of trypsin-like serine proteinases and a new functional member of the human ov-serpin family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SERPINB12
Gene Accession No. Q96P63
Gene Alias MGC119247; MGC119248; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 12; serpin B12; serpin family B member 12; serpin peptidase inhibitor, clade B (ovalbumin), member 12; SERPINB12; YUKOPIN
Gene Symbols SERPINB12
Host Species Rabbit
Immunogen Reacts with residues 260-275 of the human SERPINB12 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.