Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB13 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene
SDP

Catalog No. 502540767
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0767 30 μL
50-167-4514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0767 Supplier OriGene Supplier No. TA503674S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB13 Monoclonal antibody specifically detects SERPINB13 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen SERPINB13
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1B12
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINB13
Gene Accession No. Q9UIV8
Gene Alias 5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINB13 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5275
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.