Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB13 Monoclonal Antibody (OTI2B6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503672S


Description
SERPINB13 Monoclonal antibody specifically detects SERPINB13 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
SERPINB13 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8IYX1 | |
Mouse | |
Affinity chromatography | |
RUO | |
161514 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B6 | |
Unconjugated | |
TBC1D21 | |
5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7 | |
Full length human recombinant protein of human TBC1D21 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction