Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SERPINB13 |
---|---|
Concentration | 0.38 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1957
![]() ![]() |
Proteintech Group Inc
180451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is underexpressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.Specifications
SERPINB13 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8CDC0, Q9UIV8 | |
241196, 304690, 5275 | |
SERPINB13 Fusion Protein Ag12686 | |
Primary | |
-20°C | |
SERPINB13 |
0.38 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HaCaT UV repressible serpin, headpin, HSHUR7SEQ, HUR7, Hurpin, Peptidase inhibitor 13, PI 13, PI13, Proteinase inhibitor 13, Serpin B13, SERPINB13 | |
SERPINB13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title