Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SerpinB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177964
Description
SerpinB2 Polyclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
SerpinB2 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
Rabbit | |
46.6 kDa | |
0.1 mg | |
Apoptosis, Cancer | |
5055 | |
Human, Primate | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
1.2 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:100000-150000, Immunohistochemistry 10ug/ml | |
NP_002566.1 | |
SERPINB2 | |
This affinity purified SerpinB2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to an internal area of human SerpinB2 protein. (Uniprot: P05120) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity-purified antibody is directed against human SerpinB2 protein. Reactivity occurs against human SerpinB2 protein. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction