Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SerpinB2 Antibody (OTI1G3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274131FR
Description
SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
SerpinB2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
SERPINB2 | |
Full length recombinant protein of human SerpinB2 (NP_002566) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1G3 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer | |
5055.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction