Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SerpinB2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317002100UL
Description
SerpinB2 Polyclonal antibody specifically detects SerpinB2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SerpinB2 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MVYMGSRGSTEDQMAKVLQFNEVGANAVTPMTPENFTSCGFMQQIQKGSYPDAILQAQAADKIHSSFRSLSSAINASTGNYLLESV | |
| 100 μg | |
| Apoptosis, Cancer | |
| 5055 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction