Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SERPINB2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0664
![]() ![]() |
Proteintech Group Inc
160351AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SERPINB2 is also known as PAI or PAI2. Plasminogen activator inhibitor-2 (placental PAI) is acoagulation factor that inactivates tPA and urokinase. It is present in most cells, especially monocytes/macrophages. PAI-2 exists in two forms, a 60-kDa extracellular glycosylated form and a 43-kDa intracellular form. It is present only at detectable quantities in blood during pregcy, as it is produced by the placenta, and may explain partially the increased rate of thrombosisduring pregcy. The majority of expressed PAI-2 remains unsecreted due to the presence of an inefficient internal signal peptide.Specifications
SERPINB2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P05120 | |
5055 | |
SERPINB2 Fusion Protein Ag8906 | |
Primary | |
-20°C | |
SERPINB2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HsT1201, Monocyte Arg serpin, PAI, PAI 2, PAI2, PAI-2, PLANH2, Serpin B2, SERPIN2, SERPINB2, Urokinase inhibitor | |
SERPINB2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title