Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB3 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $437.00
Specifications
Antigen | SERPINB3 |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550936
|
Invitrogen™
MA550936 |
20 μL |
Each for $195.80
|
|
|||||
FEMA550935
|
Invitrogen™
MA550935 |
200 μL |
Each for $437.00
|
|
|||||
Description
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt]Specifications
SERPINB3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
SERPINB3 | |
IgG2b κ | |
Protein A/G | |
SERPINB3 |
C1 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P29508 | |
6317 | |
Recombinant Human SERPINB3 protein, Met1-Ile210 (Accession #Q9BYH9). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title