Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB3 Recombinant Rabbit Monoclonal Antibody (018)
GREENER_CHOICE

Catalog No. PIMA529557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529557 Supplier Invitrogen™ Supplier No. MA529557
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human SERPINB3/SCCA-1.

May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB3
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone 018
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SERPINB3
Gene Accession No. P29508
Gene Alias HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A
Gene Symbols SERPINB3
Host Species Rabbit
Immunogen Recombinant Human SERPINB3/SCCA-1 Protein (Asn2-Pro390).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.