Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB3 Recombinant Rabbit Monoclonal Antibody (6J3U6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542597
This item is not returnable.
View return policy
Description
The target is usually found in the following locations: Cytoplasm. Immunogen sequence: TMGMVDIFNG DADLSGMTGS RGLVLSGVLH KAFVEVTEEG AEAAAATAVV GFGSSPTSTN EEFHCNHPFL FFIRQNKTNS ILFYGRFSSP.
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt].
Specifications
SERPINB3 | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29508 | |
SERPINB3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 301-390 of human SSCA1/SerpinB3 (P29508). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
6J3U6 | |
Unconjugated | |
SERPINB3 | |
HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
304688, 6317 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction