Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB3 Recombinant Rabbit Monoclonal Antibody (JE55-93)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542502
This item is not returnable.
View return policy
Description
Positive Control: A431 cell lysates, human skin tissue, human esophagus tissue. Subcellular Location: Cytoplasm.
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt].
Specifications
SERPINB3 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P29508 | |
SERPINB3 | |
Recombinant protein within N-terminal Human SerpinB3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
JE55-93 | |
Unconjugated | |
SERPINB3 | |
HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
Rabbit | |
Protein A | |
RUO | |
6317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction