Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB3 + SERPINB4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988910
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8910 Supplier Bioss Supplier No. BS9214R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Metastasis of a primary tumor to a distant site is determined through signaling cascades that break down interactions between the cell and extracellular matrix proteins. Among the proteins mediating metastasis are serine prote-ases, such as neutrophil elastase. In 1985, Dr. Jim Travis and Dr. R.W. Carrell designated an emerging family of serine protease inhibitors as the serpin fam-ily, which share homology in both primary amino acid sequence and tertiary structure. Serpins contain a stretch of peptide that mimics a true substrate for a corresponding serine protease. Serine proteases bind to this substrate mimic in a 1:1 stoichiometric fashion and become catalytically inactive. Aberrant ex-pression of serpin family members can contribute to a number of conditions, including emphysema (a-1 antitrypsin deficiency), fatal bleeding (elastase to thrombin specificity) and thrombosis (antithrombin deficiency), and are indicators of cancer stage phenotypes (circulating levels of squamous cell carcinoma antigen, known as SCCA1, increase in advancing stages of some cervical, lung, esophageal and head and neck cancers). Human chromosome position 18q21.3 contains a cluster of serpins, including a tandem duplication of the SCCA gene, plasminogen activator inhibitor type 2, and maspin. SCCA is transcribed by two nearly identical genes (SCCA1 and SCCA2), and is mainly produced as SCCA1. The human SCCA1 gene encodes a 390 amino acid protein that was originally isolated from a metastatic cervical squamous cell carcinoma.

Specifications

Antigen SERPINB3 + SERPINB4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SERPINB4
Gene Accession No. P29508, P48594
Gene Alias 1110001H02Rik; 1110013A16Rik; HsT1196; Leupin; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); Protein T4-A; RGD1562868; SCC; SCCA; SCCA1; SCCA-1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; SCCA-PD; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serine protease inhibitor B3; Serpin B3; Serpin B4; serpin family B member 3; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; Serpinb3a; SERPINB4; Sqn5; Squamous cell carcinoma antigen 1; Squamous cell carcinoma antigen 2; SSCA1; T4-A
Gene Symbols Serpinb3, SERPINB4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SERPINB3 (same as SERPINB4).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6317, 6318
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.