Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB4 Antibody (001), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290154AF750
Description
SERPINB4 Monoclonal specifically detects SERPINB4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SERPINB4 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SERPINB4 (P48594; Met1-Pro390). | |
0.1 mL | |
Immunology | |
6318 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction