Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB4 Antibody (8B8B2), Alexa Fluor™ 532 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305885AF532
Description
SERPINB4 Monoclonal antibody specifically detects SERPINB4 in Human samples. It is validated for ELISASpecifications
SERPINB4 | |
Monoclonal | |
Alexa Fluor 532 | |
Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SERPINB4 (P48594; Met 1-Pro 390). | |
0.1 mL | |
Immunology | |
6318 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
8B8B2 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction