Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB4 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300059APC
Description
SERPINB4 Polyclonal antibody specifically detects SERPINB4 in Human samples. It is validated for ELISASpecifications
SERPINB4 | |
Polyclonal | |
ELISA | |
Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2 | |
Produced in rabbits immunized with purified, recombinant Human SERPINB4 (Accession#: P48594; Met1-Pro390) | |
0.1 mL | |
Immunology | |
6318 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction