Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB4 Antibody (OTI4C8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274141F
Description
SERPINB4 Monoclonal specifically detects SERPINB4 in Human samples. It is validated for Immunohistochemistry.Specifications
SERPINB4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SERPINB4 | |
Full length recombinant protein of human SERPINB4 (NP_002965) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI4C8 | |
Immunohistochemistry | |
Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2 | |
Mouse | |
Affinity Purified | |
Immunology | |
6318.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction