Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB4 Monoclonal Antibody (H10)

Catalog No. PIMA543585
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543585 20 μL
PIMA541675 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543585 Supplier Invitrogen™ Supplier No. MA543585
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SERPINB4 Monoclonal Antibody for Western Blot, IHC (P)

SERPINB4 is also called squamous cell carcinoma antigen 2 (SCCA-2) or leupin. It directly inhibits human granzyme proteolytic activity and that overexpression of SERPINB4 in HeLa cells inhibits recombinant granzyme-induced as well as NK cell-mediated cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SERPINB4
Gene Accession No. P48594
Gene Alias 1110001H02Rik; 1110013A16Rik; LEUPIN; Leupin SERPINB4; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); RGD1562868; SCCA1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serpin B4; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3a; SERPINB4; Sqn5; squamous cell carcinoma antigen 1; squamous cell carcinoma antigen 2
Gene Symbols SERPINB4
Host Species Mouse
Immunogen Recombinant protein Serpin Peptidase Inhibitor, Clade B, Member 4. The antigen corresponds to amino acid range 144-298 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.