Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB4 Monoclonal Antibody (OTI1G6), TrueMAB™, OriGene
SDP

Catalog No. 502541630
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1630 30 μL
50-167-3389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1630 Supplier OriGene Supplier No. TA502327S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB4 Monoclonal antibody specifically detects SERPINB4 in Human samples. It is validated for Western Blot

Specifications

Antigen SERPINB4
Applications Western Blot
Classification Monoclonal
Clone OTI1G6
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINB4
Gene Accession No. P48594
Gene Alias 1110001H02Rik; 1110013A16Rik; LEUPIN; Leupin SERPINB4; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); RGD1562868; SCCA1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serpin B4; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3a; SERPINB4; Sqn5; squamous cell carcinoma antigen 1; squamous cell carcinoma antigen 2
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINB4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6318
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.