Learn More
SERPINB4 Mouse anti-Human, Clone: OTI2B9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502323

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SERPINB4 is also called squamous cell carcinoma antigen 2 (SCCA-2) or leupin. It directly inhibits human granzyme proteolytic activity and that overexpression of SERPINB4 in HeLa cells inhibits recombit granzyme-induced as well as NK cell-mediated cell death.Specifications
SERPINB4 | |
Monoclonal | |
Unconjugated | |
SERPINB4 | |
LEUPIN, PI11, SCCA-2, SCCA1, SCCA2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6318 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2B9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P48594 | |
SERPINB4 | |
Full length human recombit protein of human SERPINB4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.