Learn More
Invitrogen™ SERPINB4 Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | SERPINB4 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| Gene | SERPINB4 |
| Gene Accession No. | P48594 |
| Gene Alias | 1110001H02Rik; 1110013A16Rik; LEUPIN; Leupin SERPINB4; Peptidase inhibitor 11; PI11; PI-11; protease inhibitor (leucine-serpin); RGD1562868; SCCA1; SCCA2; SCCA-2; SCCA2/SCCA1 fusion protein; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3A; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 4; serine peptidase inhibitor, clade B, member 3A; serpin B4; serpin family B member 4; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3a; SERPINB4; Sqn5; squamous cell carcinoma antigen 1; squamous cell carcinoma antigen 2 |
| Gene Symbols | Serpinb3a, SERPINB4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.