Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB5, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31620
Description
Sequence: TFCMGNIDSINCKIIELPFQNKHLSMFILLPKDVEDESTGLEKIEKQLNSESLSQWTNPSTMANAKVKLSIPKFKVEKMIDPKSpecifications
SERPINB5 | |
Polyclonal | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
SERPINB5 | |
Recombinant protein corresponding to amino acids 203-285 of human SERPINB5. | |
RUO | |
5268 | |
Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
PI5/maspin | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction