Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB6 Monoclonal Antibody (OTI1C5)
GREENER_CHOICE

Catalog No. PIMA525823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525823 Supplier Invitrogen™ Supplier No. MA525823
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB6 Monoclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene is a member of the serpin superfamily, and ovalbumin-serpin subfamily. It was originally discovered as a placental thrombin inhibitor. The mouse homolog was found to be expressed in the hair cells of the inner ear. Mutations in this gene are associated with nonsyndromic progressive hearing loss, suggesting that this serpin plays an important role in the inner ear in the protection against leakage of lysosomal content during stress, and that loss of this protection results in cell death and sensorineural hearing loss. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SERPINB6
Gene Accession No. P35237, Q60854
Gene Alias 4930482L21Rik; CAP; Cytoplasmic antiproteinase; D330015H01Rik; DFNB91; DKFZp686I04222; MGC111370; MSTP057; ovalbumin; Peptidase inhibitor 6; PI6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); proteinase inhibitor 6; PTI; QtsA-17114; RP1-90J20.6; serine (or cysteine) peptidase inhibitor, clade B, member 6a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; serine (or cysteine) proteinase inhibitor, clade B, member 6a; serine protease inhibitor 3; serine proteinase inhibitor B-43; Serpin B6; serpin family B member 6; serpin peptidase inhibitor, clade B (ovalbumin), member 6; Serpinb6; Serpinb6a; Spi3; thrombin inhibitor; unnamed protein product
Gene Symbols SERPINB6, Serpinb6a
Host Species Mouse
Immunogen Full length human recombinant protein of SERPINB6 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20719, 5269
Target Species Human, Mouse, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.