Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB6 Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
SDP

Catalog No. 502540532
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0532 30 μL
50-167-4825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0532 Supplier OriGene Supplier No. TA504055S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERPINB6 Monoclonal antibody specifically detects SERPINB6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SERPINB6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F8
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Enpep
Gene Accession No. Q07075
Gene Alias 4930482L21Rik; CAP; Cytoplasmic antiproteinase; D330015H01Rik; DFNB91; DKFZp686I04222; MGC111370; MSTP057; ovalbumin; Peptidase inhibitor 6; PI6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); proteinase inhibitor 6; PTI; QtsA-17114; RP1-90J20.6; serine (or cysteine) peptidase inhibitor, clade B, member 6a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; serine (or cysteine) proteinase inhibitor, clade B, member 6a; serine protease inhibitor 3; serine proteinase inhibitor B-43; Serpin B6; serpin family B member 6; serpin peptidase inhibitor, clade B (ovalbumin), member 6; Serpinb6; Serpinb6a; Spi3; thrombin inhibitor; unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human ENPEP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2028
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.