Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518947
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518947 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518947 Supplier Invitrogen™ Supplier No. PA518947
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

The protein encoded by this gene is a member of the serpin superfamily, and ovalbumin-serpin subfamily. It was originally discovered as a placental thrombin inhibitor. The mouse homolog was found to be expressed in the hair cells of the inner ear. Mutations in this gene are associated with nonsyndromic progressive hearing loss, suggesting that this serpin plays an important role in the inner ear in the protection against leakage of lysosomal content during stress, and that loss of this protection results in cell death and sensorineural hearing loss. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINB6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SERPINB6
Gene Accession No. P35237
Gene Alias 4930482L21Rik; CAP; Cytoplasmic antiproteinase; D330015H01Rik; DFNB91; DKFZp686I04222; MGC111370; MSTP057; ovalbumin; Peptidase inhibitor 6; PI6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); proteinase inhibitor 6; PTI; QtsA-17114; RP1-90J20.6; serine (or cysteine) peptidase inhibitor, clade B, member 6a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; serine (or cysteine) proteinase inhibitor, clade B, member 6a; serine protease inhibitor 3; serine proteinase inhibitor B-43; Serpin B6; serpin family B member 6; serpin peptidase inhibitor, clade B (ovalbumin), member 6; Serpinb6; Serpinb6a; Spi3; thrombin inhibitor; unnamed protein product
Gene Symbols SERPINB6
Host Species Goat
Immunogen Synthetic peptide sequence (NWDEQFDKENTEER) corresponding to the internal amino acids of SERPINB6 (aa 172-186).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5269
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.