Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SerpinB7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309378100UL
Description
SerpinB7 Polyclonal specifically detects SerpinB7 in Human samples. It is validated for Western Blot.Specifications
SerpinB7 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
DKFZp686D06190, Megsin, MEGSINclade B (ovalbumin), member 7, mesangium predominant gene, megsin, MGC120014, MGC120015, serpin B7, serpin peptidase inhibitor, clade B (ovalbumin), member 7, TP55 | |
The immunogen is a synthetic peptide directed towards the N terminal region of human SerpinB7 (NP_003775). Peptide sequence LSQIDKLLHVNTASGYGNSSNSQSGLQSQLKRVFSDINASHKDYDLSIVN | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
8710 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction