Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB8 Monoclonal Antibody (OTI9B2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810581

Description
The protein encoded by this gene is a member of the ov-serpin family of serine protease inhibitors. The encoded protein is produced by platelets and can bind to and inhibit the function of furin, a serine protease involved in platelet functions. In addition, this protein has been found to enhance the mechanical stability of cell-cell adhesion in the skin, and defects in this gene have been associated with an autosomal-recessive form of exfoliative ichthyosis.Specifications
SERPINB8 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P50452 | |
SERPINB8 | |
Human recombit protein fragment corresponding to amino acids 169-374 of human SERPINB8 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI9B2 | |
Unconjugated | |
SERPINB8 | |
C18orf53, CAP2, HMSD, HSMD-v, PI-8, PI8, PSS5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5271 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction